SY3H3D6821FC Synthonix Corporation

N-Isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number No MDL found
  • Cas Number : 1256360-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58477.html

Safety Information

MSDS https://synthonix.com/58477.html
Availability details for
Quantity
  • Availability
  • Add to Cart