SY3H3D6821FD Synthonix Corporation

1-(Isopropylamino)-3-(2-(2-methoxyethyl)phenoxy)propan-2-ol

  • MDL number MFCD18379554
  • Cas Number : 163685-38-9
  • Molecular Weight : 267.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/58488.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/58488.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart