SY3H3D682200 Synthonix Corporation

Pentadecanedioic acid

  • MDL number No MDL found
  • Cas Number : 1460-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/58506.html

Safety Information

MSDS https://synthonix.com/58506.html
Availability details for
Quantity
  • Availability
  • Add to Cart