SY3H3D682201 Synthonix Corporation

Isopropyl dodecanoate

  • MDL number No MDL found
  • Cas Number : 10233-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/58510.html

Safety Information

MSDS https://synthonix.com/58510.html
Availability details for
Quantity
  • Availability
  • Add to Cart