SY3H3D682203 Synthonix Corporation

4-(2-Phenylpropan-2-yl)aniline hydrochloride

  • MDL number No MDL found
  • Cas Number : 1416354-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58535.html

Safety Information

MSDS https://synthonix.com/58535.html
Availability details for
Quantity
  • Availability
  • Add to Cart