SY3H3D682205 Synthonix Corporation

4,4'-(Propane-1,3-diylbis(oxy))bis(benzene-1,3-diamine) tetrahydrochloride

  • MDL number No MDL found
  • Cas Number : 74918-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58545.html

Safety Information

MSDS https://synthonix.com/58545.html
Availability details for
Quantity
  • Availability
  • Add to Cart