SY3H3D682209 Synthonix Corporation

Perfluorophenyl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate

  • Cas Number : 120550-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58648.html

Safety Information

MSDS https://synthonix.com/58648.html
Availability details for
Quantity
  • Availability
  • Add to Cart