SY3H3D68220A Synthonix Corporation

4'-Propoxy-[1,1'-biphenyl]-4-carbonitrile

  • MDL number No MDL found
  • Cas Number : 52709-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58652.html

Safety Information

MSDS https://synthonix.com/58652.html
Availability details for
Quantity
  • Availability
  • Add to Cart