SY3H3D68220C Synthonix Corporation

4-(trans-4-((E)-Prop-1-en-1-yl)cyclohexyl)benzonitrile

  • MDL number No MDL found
  • Cas Number : 96184-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58717.html

Safety Information

MSDS https://synthonix.com/58717.html
Availability details for
Quantity
  • Availability
  • Add to Cart