SY3H3D68220D Synthonix Corporation

1-(5-Phenylpyridin-3-yl)-1,4-diazepane

  • MDL number No MDL found
  • Cas Number : 223796-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/58719.html

Safety Information

MSDS https://synthonix.com/58719.html
Availability details for
Quantity
  • Availability
  • Add to Cart