SY3H3D68220F Synthonix Corporation

4-(trans-4-propylcyclohexyl)benzonitrile

  • Cas Number : 61203-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58750.html

Safety Information

MSDS https://synthonix.com/58750.html
Availability details for
Quantity
  • Availability
  • Add to Cart