SY3H3D682211 Synthonix Corporation

1-Phenyl-2,5,8,11-tetraoxatridecan-13-yl methanesulfonate

  • MDL number No MDL found
  • Cas Number : 477781-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58846.html

Safety Information

MSDS https://synthonix.com/58846.html
Availability details for
Quantity
  • Availability
  • Add to Cart