SY3H3D682212 Synthonix Corporation

4,4'-(Propane-2,2-diyl)bis(cyanatobenzene)

  • MDL number No MDL found
  • Cas Number : 1156-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58904.html

Safety Information

MSDS https://synthonix.com/58904.html
Availability details for
Quantity
  • Availability
  • Add to Cart