SY3H3D682215 Synthonix Corporation

2-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number No MDL found
  • Cas Number : 1313519-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58996.html

Safety Information

MSDS https://synthonix.com/58996.html
Availability details for
Quantity
  • Availability
  • Add to Cart