SY3H3D682217 Synthonix Corporation

2-Phenoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 330792-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59000.html

Safety Information

MSDS https://synthonix.com/59000.html
Availability details for
Quantity
  • Availability
  • Add to Cart