SY3H3D68221A Synthonix Corporation

(4'-(Pentyloxy)-[1,1'-biphenyl]-4-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 158937-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59020.html

Safety Information

MSDS https://synthonix.com/59020.html
Availability details for
Quantity
  • Availability
  • Add to Cart