SY3H3D68221B Synthonix Corporation

4-(trans-4-Pentylcyclohexyl)phenol

  • Cas Number : 82575-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/59079.html

Safety Information

MSDS https://synthonix.com/59079.html
Availability details for
Quantity
  • Availability
  • Add to Cart