SY3H3D68221E Synthonix Corporation

(4R,4'R)-2,2'-(Propane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

  • MDL number No MDL found
  • Cas Number : 131833-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59093.html

Safety Information

MSDS https://synthonix.com/59093.html
Availability details for
Quantity
  • Availability
  • Add to Cart