SY3H3D68221F Synthonix Corporation

4-(trans-4-Pentylcyclohexyl)cyclohexanone

  • Cas Number : 84868-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59095.html

Safety Information

MSDS https://synthonix.com/59095.html
Availability details for
Quantity
  • Availability
  • Add to Cart