SY3H3D682220 Synthonix Corporation

N-((2E,4E)-5-(Phenylamino)penta-2,4-dien-1-ylidene)aniline hydrochloride

  • MDL number MFCD00012630
  • Cas Number : 1497-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Brown - Red - Deep purple
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59102.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59102.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart