SY3H3D682222 Synthonix Corporation

Potassium heptadecanoate

  • MDL number No MDL found
  • Cas Number : 17378-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59112.html

Safety Information

MSDS https://synthonix.com/59112.html
Availability details for
Quantity
  • Availability
  • Add to Cart