SY3H3D682226 Synthonix Corporation

3-Phenyl-9H-carbazole

  • MDL number No MDL found
  • Cas Number : 103012-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/59156.html

Safety Information

MSDS https://synthonix.com/59156.html
Availability details for
Quantity
  • Availability
  • Add to Cart