SY3H3D682228 Synthonix Corporation

(9-Phenyl-9H-carbazol-2-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 1001911-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/59169.html

Safety Information

MSDS https://synthonix.com/59169.html
Availability details for
Quantity
  • Availability
  • Add to Cart