SY3H3D68222C Synthonix Corporation

Piperazine-1,4-diylbis((4-chlorophenyl)methanone)

  • MDL number No MDL found
  • Cas Number : 107785-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59187.html

Safety Information

MSDS https://synthonix.com/59187.html
Availability details for
Quantity
  • Availability
  • Add to Cart