SY3H3D68222E Synthonix Corporation

N,N'-(1,3-Phenylenebis(methylene))bis(furan-2-carboxamide)

  • MDL number No MDL found
  • Cas Number : 415694-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/59190.html

Safety Information

MSDS https://synthonix.com/59190.html
Availability details for
Quantity
  • Availability
  • Add to Cart