SY3H3D68222F Synthonix Corporation

4'-Pentyl-[1,1'-biphenyl]-4-carboxylic acid

  • Cas Number : 59662-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59237.html

Safety Information

MSDS https://synthonix.com/59237.html
Availability details for
Quantity
  • Availability
  • Add to Cart