SY3H3D682231 Synthonix Corporation

2-((((1S,2R,5S)-2-Isopropyl-5-methylcyclohexyl)oxy)carbonyl)benzoic acid

  • Cas Number : 53623-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59283.html

Safety Information

MSDS https://synthonix.com/59283.html
Availability details for
Quantity
  • Availability
  • Add to Cart