SY3H3D682234 Synthonix Corporation

(2S,5R)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 5-(4-amino-5-fluoro-2-oxopyrimidin-1(2H)-yl)-1,3-oxathiolane-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 147126-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59304.html

Safety Information

MSDS https://synthonix.com/59304.html
Availability details for
Quantity
  • Availability
  • Add to Cart