SY3H3D682236 Synthonix Corporation

Isopentyl pentyl phthalate

  • MDL number No MDL found
  • Cas Number : 776297-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59310.html

Safety Information

MSDS https://synthonix.com/59310.html
Availability details for
Quantity
  • Availability
  • Add to Cart