SY3H3D682239 Synthonix Corporation

4,4'-(Pyridine-2,5-diyl)dibenzoic acid

  • Cas Number : 1800425-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59354.html

Safety Information

MSDS https://synthonix.com/59354.html
Availability details for
Quantity
  • Availability
  • Add to Cart