SY3H3D68223F Synthonix Corporation

4,4'-(Perfluoropropane-2,2-diyl)bis(1,2-dimethylbenzene)

  • MDL number MFCD00042167
  • Cas Number : 65294-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59388.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59388.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart