SY3H3D682242 Synthonix Corporation

(Propane-2,2-diylbis(2-hydroxybenzene-5,3,1-triyl))tetramethanol

  • MDL number MFCD00071732
  • Cas Number : 3957-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59429.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/59429.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart