SY3H3D682243 Synthonix Corporation

1-(trans-4-Pentylcyclohexyl)-4-ethoxybenzene

  • MDL number No MDL found
  • Cas Number : 84540-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59456.html

Safety Information

MSDS https://synthonix.com/59456.html
Availability details for
Quantity
  • Availability
  • Add to Cart