SY3H3D682249 Synthonix Corporation

(S)-5-Phenyl-1H-pyrrol-3-yl 2-(4-methylphenylsulfonamido)propanoate

  • MDL number No MDL found
  • Cas Number : 99740-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59535.html

Safety Information

MSDS https://synthonix.com/59535.html
Availability details for
Quantity
  • Availability
  • Add to Cart