SY3H3D68224A Synthonix Corporation

3-Phenyl-1-(tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

  • MDL number No MDL found
  • Cas Number : 1028092-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59579.html

Safety Information

MSDS https://synthonix.com/59579.html
Availability details for
Quantity
  • Availability
  • Add to Cart