SY3H3D68224B Synthonix Corporation

8-(Phenylmethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-8-azabicyclo[3.2.1]oct-2-ene

  • MDL number No MDL found
  • Cas Number : 1123661-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59588.html

Safety Information

MSDS https://synthonix.com/59588.html
Availability details for
Quantity
  • Availability
  • Add to Cart