SY3H3D68224F Synthonix Corporation

Quinazolin-8-ol

  • MDL number No MDL found
  • Cas Number : 7557-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/47864.html

Safety Information

MSDS https://synthonix.com/47864.html
Availability details for
Quantity
  • Availability
  • Add to Cart