SY3H3D682251 Synthonix Corporation

Quinoxaline-2,3-diamine

  • MDL number No MDL found
  • Cas Number : 6640-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48862.html

Safety Information

MSDS https://synthonix.com/48862.html
Availability details for
Quantity
  • Availability
  • Add to Cart