SY3H3D682253 Synthonix Corporation

Quinuclidin-4-ylmethanamine dihydrochloride

  • Cas Number : 67496-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49679.html

Safety Information

MSDS https://synthonix.com/49679.html
Availability details for
Quantity
  • Availability
  • Add to Cart