SY3H3D68225D Synthonix Corporation

Quinolin-7-amine hydrochloride

  • Cas Number : 1246556-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52219.html

Safety Information

MSDS https://synthonix.com/52219.html
Availability details for
Quantity
  • Availability
  • Add to Cart