SY3H3D682264 Synthonix Corporation

Isoquinolin-3-ylmethanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1628557-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/54084.html

Safety Information

MSDS https://synthonix.com/54084.html
Availability details for
Quantity
  • Availability
  • Add to Cart