SY3H3D682269 Synthonix Corporation

2-(Quinolin-6-yl)acetic acid

  • Cas Number : 5622-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55357.html

Safety Information

MSDS https://synthonix.com/55357.html
Availability details for
Quantity
  • Availability
  • Add to Cart