SY3H3D68226D Synthonix Corporation

(S)-N-((S)-Quinuclidin-3-yl)-1,2,3,4-tetrahydronaphthalene-1-carboxamide

  • MDL number No MDL found
  • Cas Number : 177793-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59278.html

Safety Information

MSDS https://synthonix.com/59278.html
Availability details for
Quantity
  • Availability
  • Add to Cart