SY3H3D682272 Synthonix Corporation

Sodium 3-(cyclohexylamino)propane-1-sulfonate

  • MDL number No MDL found
  • Cas Number : 105140-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39557.html

Safety Information

MSDS https://synthonix.com/39557.html
Availability details for
Quantity
  • Availability
  • Add to Cart