SY3H3D68227E Synthonix Corporation

Sodium oxazole-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1255098-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40296.html

Safety Information

MSDS https://synthonix.com/40296.html
Availability details for
Quantity
  • Availability
  • Add to Cart