SY3H3D682282 Synthonix Corporation

Sodium 5-formylfuran-2-sulfonate

  • MDL number No MDL found
  • Cas Number : 31795-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40691.html

Safety Information

MSDS https://synthonix.com/40691.html
Availability details for
Quantity
  • Availability
  • Add to Cart