SY3H3D682284 Synthonix Corporation

Sodium 3-((2-aminoethyl)amino)propanoate

  • MDL number No MDL found
  • Cas Number : 84434-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/41302.html

Safety Information

MSDS https://synthonix.com/41302.html
Availability details for
Quantity
  • Availability
  • Add to Cart