SY3H3D68228D Synthonix Corporation

Sodium 4-hydroxyphenyl phosphate

  • MDL number No MDL found
  • Cas Number : 20368-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42720.html

Safety Information

MSDS https://synthonix.com/42720.html
Availability details for
Quantity
  • Availability
  • Add to Cart