SY3H3D68228F Synthonix Corporation

Sodium 1-cyano-3-ethoxy-2,3-dioxopropan-1-ide

  • Cas Number : 53544-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43425.html

Safety Information

MSDS https://synthonix.com/43425.html
Availability details for
Quantity
  • Availability
  • Add to Cart