SY3H3D682290 Synthonix Corporation

3-Sulfopropyl Methacrylate Potassium Salt

  • MDL number No MDL found
  • Cas Number : 31098-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43587.html

Safety Information

MSDS https://synthonix.com/43587.html
Availability details for
Quantity
  • Availability
  • Add to Cart